Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDCA3 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10074621UV
Description
CDCA3 Polyclonal antibody specifically detects CDCA3 in Human samples. It is validated for Western BlotSpecifications
CDCA3 | |
Polyclonal | |
Western Blot | |
C8, cell division cycle associated 3, FBXO52, Gene-rich cluster protein C8, GRCC8cell division cycle-associated protein 3, MGC2577, TOME1, TOME-1gene rich cluster, C8, trigger of mitotic entry 1, Trigger of mitotic entry protein 1 | |
The immunogen recognized by this antibody maps to a region between residue 200 and 230 of human cell division cycle associated 3 (trigger of mitotic entry 1) using the numbering given in entry NP_112589.1 (GeneID 83461). | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
83461 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction