Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDCA3 Monoclonal Antibody (OTI6F7), TrueMAB™, OriGene
SDP

Catalog No. 502543940
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3940 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3940 Supplier OriGene Supplier No. CF811694
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CDCA3 Monoclonal antibody specifically detects CDCA3 in Human samples. It is validated for Western Blot

Specifications

Antigen CDCA3
Applications Western Blot
Classification Monoclonal
Clone OTI6F7
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene E2f4
Gene Accession No. Q16254
Gene Alias 2410005A12Rik; C8; CDCA 3; CDCA3; cell division cycle associated 3; cell division cycle-associated protein 3; gene rich cluster, C8; gene-rich cluster protein C8; GRCC 8; GRCC8; MGC2577; TOME 1; Tome1; TOME-1; trigger of mitotic entry 1; trigger of mitotic entry protein 1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 86-413 of human E2F4 produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1874
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.