Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDCA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110918
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110918 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110918 Supplier Invitrogen™ Supplier No. PA5110918
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KEEARQPTET PVASQSSDKP SRDPETPRSS GSMRNRWKPN SSKVLGRSPL TILQDDNSPG TLTLRQGKRP SPLSENVSEL KEGAILGTGR LLKTGGRAWE Q Highest antigen sequence indentity to the following orthologs: Mouse - 78%, Rat - 80%.

F-box-like protein which is required for entry into mitosis. Acts by participating in E3 ligase complexes that mediate the ubiquitination and degradation of WEE1 kinase at G2/M phase. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDCA3
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDCA3
Gene Accession No. Q99618
Gene Alias 2410005A12Rik; C8; CDCA 3; CDCA3; cell division cycle associated 3; cell division cycle-associated protein 3; gene rich cluster, C8; gene-rich cluster protein C8; GRCC 8; GRCC8; MGC2577; TOME 1; Tome1; TOME-1; trigger of mitotic entry 1; trigger of mitotic entry protein 1
Gene Symbols CDCA3
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human CDCA3. Recombinant protein control fragment (Product #RP-88748).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83461
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.