Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDCA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554867
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554867 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554867 Supplier Invitrogen™ Supplier No. PA554867
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SILPEIWPKT PSAAAVRKPI VLKRIVAHAV EVPAVQSPRR SPRISFFLEK ENEPPGRELT KEDLFKTHSV PATPTSTPVP NPEAESSSKE GELDARDLEM SKKVR Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 64%.

CDCA5 is the regulator of sister chromatid cohesion in mitosis. It may act by regulating the ability of the cohesin complex to mediate sister chromatid cohesion, perhaps by altering the nature of the interaction of cohesin with the chromosomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDCA5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDCA5
Gene Accession No. Q96FF9
Gene Alias 2610036L13Rik; AL024086; AW536684; C85404; Cdca5; cell division cycle associated 5; cell division cycle-associated protein 5; p35; RGD1560863; sororin; sororin p35
Gene Symbols CDCA5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CDCA5. Recombinant protein control fragment (Product #RP-88830).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113130
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.