Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDCA7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545892
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545892 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545892 Supplier Invitrogen™ Supplier No. PA545892
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: CRTRSQCRHS GPLRVAMKFP ARSTRGATNK KAESRQPSEN SVTDSNSDSE Sequence homology: Human: 100%.

This gene was identified as a c-Myc responsive gene, and behaves as a direct c-Myc target gene. Overexpression of this gene is found to enhance the transformation of lymphoblastoid cells, and it complements a transformation-defective Myc Box II mutant, suggesting its involvement in c-Myc-mediated cell transformation. Two alternatively spliced transcript variants encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDCA7
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CDCA7
Gene Accession No. Q9BWT1
Gene Alias 2310021G01Rik; AU044109; CDCA7; cell division cycle associated 7; cell division cycle-associated protein 7; c-Myc target JPO1; ICF3; JPO1; Protein JPO1
Gene Symbols CDCA7
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human CDCA7 (aa 58-107).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83879
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.