Learn More
CDCA7L Mouse anti-Human, Clone: OTI6C6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF802969

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Cell division cycle-associated 7-like protein (or RAM2) is a transcription factor which plays a role in transcriptional regulation and apoptotic signaling pathways. It acts as a repressor that inhibits monoamine oxidase A (MAOA) activity and gene expression by binding to promoter. It also plays an important oncogenic role in mediating the full transforming effect of MYC in medulloblastoma cells. It may act downstream of P38-kinase and BCL-2, but upstream of CASP3/caspase-3 as well as CCND1/cyclin D1 and E2F1.Specifications
CDCA7L | |
Monoclonal | |
Unconjugated | |
CDCA7L | |
JPO2, R1, RAM2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
55536 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI6C6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96GN5 | |
CDCA7L | |
Human recombit protein fragment corresponding to amino acids 1-337 of human CDCA7L produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.