Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDCA7L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555533
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555533 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555533 Supplier Invitrogen™ Supplier No. PA555533
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VRFHSKYFTE ELRRIFIEDT DSETEDFAGF TQSDLNGKTN PEVMVVESDL SDDGKASLVS EEEEDEEEDK ATPRRSRSRR SSIGLRVAFQ FPTKKLANKP DKNSSSEQLF SSARLQNEKK TILERKKDCR QVIQREDSTS ESE Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 57%.

Cell division cycle-associated 7-like protein (or RAM2) is a transcription factor which plays a role in transcriptional regulation and apoptotic signaling pathways. It acts as a repressor that inhibits monoamine oxidase A (MAOA) activity and gene expression by binding to promoter. It also plays an important oncogenic role in mediating the full transforming effect of MYC in medulloblastoma cells. It may act downstream of P38-kinase and BCL-2, but upstream of CASP3/caspase-3 as well as CCND1/cyclin D1 and E2F1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDCA7L
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDCA7L
Gene Accession No. Q96GN5
Gene Alias BC006933; CDCA7L; cell division cycle associated 7 like; cell division cycle associated 7-like; cell division cycle-associated 7-like protein; HR1; JPO2; Protein JPO2; R1; RAM2; transcription factor RAM2
Gene Symbols CDCA7L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CDCA7L. Recombinant protein control fragment (Product #RP-88699).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55536
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.