Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDCA8 Monoclonal Antibody (181CT41.2.3)
GREENER_CHOICE

Catalog No. PIMA537490
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537490 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537490 Supplier Invitrogen™ Supplier No. MA537490
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CDCA8 Monoclonal Antibody for Western Blot

CDCA8 is a component of a chromosomal passenger complex (CPC) required for stability of the bipolar mitotic spindle. The chromosomal passenger complex, which includes Survivin, CDCA8, INCENP and Aurora-B, is known to play crucial roles during mitosis and cell division. It was found that CDCA8 interacting with Aurora-B, INCENP and Survivin, increases during G2/M phase and then reduces after exit from M phase. CDCA8 is cell cycle regulated, down-regulated in response to p53/Rb-signaling, and up-regulated in many types of cancerous tissues. In Drosophila cells, inactivation of CDCA8 results in polyploidy, delayed mitosis and abnormal tissue development, indicating its critical role for cell proliferation. Recent studies show that CDCA8 is essential for cell proliferation during early embryonic development, and its early embryonic lethality cannot be rescued by the loss of p53. Its aberrant expression is linked to a poor prognosis for gastric cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDCA8
Applications Western Blot
Classification Monoclonal
Clone 181CT41.2.3
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CDCA8
Gene Accession No. Q53HL2
Gene Alias 4831429J16Rik; AU044747; BOR; Borealin; Cdca8; cell division cycle associated 8; cell division cycle-associated protein 8; D4Ertd421e; Dasra B; DasraB; dasra-B; embryonic stem cell-related; hDasra-B; MESrg; MESRGP; PESCRG3; Pluripotent embryonic stem cell-related gene 3 protein
Gene Symbols CDCA8
Host Species Mouse
Immunogen CDCA8 recombinant protein.
Purification Method Protein G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55143
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.