Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDCP1 (CD318) Monoclonal Antibody (9A2), Functional Grade, In Vivo GOLD™, Leinco Technologies™
SDP

Catalog No. 502544190
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4190 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4190 Supplier Leinco Technologies Inc Supplier No. C91005MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specificity: Clone 9A2 recognizes an epitope within the extracellular domain of human/mouse CDCP1. Endotoxin Level: <1.0 EU/mg as determined by the LAL method. Purity: ≥95% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80C. Avoid Repeated Freeze Thaw Cycles.

CDCP1 (CD318) Monoclonal antibody specifically detects CDCP1 (CD318) in Human, Mouse samples. It is validated for Flow Cytometry, Functional Assay

Specifications

Antigen CDCP1 (CD318)
Applications Flow Cytometry, Functional Assay
Classification Monoclonal
Clone 9A2
Concentration ≥5.0 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2-7.4
Gene Cdcp1
Gene Accession No. Q5U462, Q9H5V8
Gene Alias 9030022E12Rik; AA409659; CD318; CDCP1; CUB domain containing protein 1; CUB domain-containing protein 1; E030027H19Rik; gp140; Membrane glycoprotein gp140; RGD1305578; SIMA135; Subtractive immunization M plus HEp3-associated 135 kDa protein; transmembrane and associated with src kinases; TRASK; UNQ2486/PRO5773
Host Species Mouse
Immunogen His-tagged recombinant fragment from the internal region of human CUB domain-containing protein 1
Purification Method Protein A/G
Quantity 5 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109332, 64866
Target Species Human, Mouse
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.