Learn More
Invitrogen™ CDCP1 Monoclonal Antibody (309137)

Description
This antibody recognizes an epitope found between aa 33-369 of human CDCP1. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | CDCP1 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 309137 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | Cdcp1 |
| Gene Accession No. | Q9H5V8 |
| Gene Alias | 9030022E12Rik; AA409659; CD318; CDCP1; CUB domain containing protein 1; CUB domain-containing protein 1; E030027H19Rik; gp140; Membrane glycoprotein gp140; RGD1305578; SIMA135; Subtractive immunization M plus HEp3-associated 135 kDa protein; transmembrane and associated with src kinases; TRASK; UNQ2486/PRO5773 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.