Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CDCP1 Recombinant Human Monoclonal Antibody (30D4)
Human Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA550097
Description
CDCP1 Recombinant Monoclonal Antibody for ELISA
CDCP1 is a transmembrane protein containing three extracellular CUB domains. It is found to be overexpressed in colon and lung cancers. Its expression level is correlated with the metastatic ability of carcinoma cells. CDCP1 is located on the cell surface. It has been shown to be tyrosine phosphorylated in a cancer cell line. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications
CDCP1 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
Q5U462, Q9H5V8 | |
Cdcp1 | |
Recombinant Human CDCP1 protein. | |
100 μL | |
Primary | |
Human, Mouse, Monkey | |
Antibody | |
IgG1 |
ELISA, Neutralization | |
30D4 | |
Unconjugated | |
Cdcp1 | |
9030022E12Rik; AA409659; CD318; CDCP1; CUB domain containing protein 1; CUB domain-containing protein 1; E030027H19Rik; gp140; Membrane glycoprotein gp140; RGD1305578; SIMA135; Subtractive immunization M plus HEp3-associated 135 kDa protein; transmembrane and associated with src kinases; TRASK; UNQ2486/PRO5773 | |
Human | |
Affinity chromatography | |
RUO | |
109332, 64866 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction