Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CDDO Im
GREENER_CHOICESDP

Catalog No. 473710 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-371-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 47-371-0 Supplier Tocris Bioscience™ Supplier No. 4737/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Nrf2 signaling activator

CDDO Im is an Nrf2 signaling activator; increases Nrf2 protein expression and enhances Nrf2-dependent cytoprotective gene expression. CDDO Im inhibits cell growth and induces apoptosis of pancreatic cancer cells in vitro. Also reduces acetaminophen -induced liver injury in mice.

Specifications

Quantity 10 mg
Product Type CDDO Im
Purity >98%
Molecular Weight (g/mol) 541.72

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.