Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDH1 Mouse anti-Human, Clone: 4A2, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB15076
Description
Mouse monoclonal antibody raised against full length recombinant human CDH1.
- High specificity
- High reactivity
Specifications
CDH1 | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined | |
P12830 | |
CDH1 | |
Recombinant protein corresponding to full length human CDH1. | |
100 μg | |
Primary | |
Human | |
Liquid |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
4A2 | |
Mouse monoclonal antibody raised against full length recombinant human CDH1. | |
In 10 mM PBS (0.05% BSA, 0.05% sodium azide). | |
Arc-1/CD324/CDHE/ECAD/LCAM/UVO | |
Mouse | |
Protein A/G purification | |
RUO | |
999 | |
Store at 4°C. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction