Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDH1 Rabbit anti-Human, Mouse, Polyclonal, Abnova™

Catalog No. 89035700 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
40 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-700 40 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-700 Supplier Abnova Corporation Supplier No. PAB30207

Rabbit Polyclonal Antibody

Rabbit polyclonal antibody raised against synthetic peptide of human CDH1.

  • High specificity
  • High reactivity

Specifications

Antigen CDH1
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of human CDH1.
Dilution Flow Cytometry (1-3 ug x 106 cells) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS, pH 7.4 (0.02% sodium azide).
Gene Accession No. P12830
Gene Alias Arc-1/CD324/CDHE/ECAD/LCAM/UVO
Gene Symbols CDH1
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 650-700 of human CDH1.
Purification Method Immunoaffinity purification
Quantity 40 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 999
Target Species Human, Mouse
Content And Storage Store at -20°C on dry atmosphere, lyophilized antibodies are stable.
After reconstitution with deionized water, store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.