Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDH1 Rabbit anti-Human, Unconjugated, Clone: CDH1/2208R, Abnova™

Catalog No. 89339724 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-724 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-724 Supplier Abnova Corporation Supplier No. RAB00685
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

CDH1 Monoclonal antibody specifically detects CDH1 in Human samples. It is validated for WB-Ce, IHC-P, IF, Flow Cyt.

Specifications

Antigen CDH1
Applications Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone CDH1/2208R
Conjugate Unconjugated
Description cadherin 1, type 1, E-cadherin (epithelial)
Gene CDH1
Gene Alias Arc-1, CD324, CDHE, ECAD, LCAM, UVO
Host Species Rabbit
Immunogen Recombinant protein corresponding to full-length human CDH1
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 999
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.