Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CDH10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144705
Description
Immunogen sequence: SGTTTVNITL TDVNDNPPRF PQNTIHLRVL ESSPVGTAIG SVKATDADTG KNAEVEYRII DGDGTDMFDI VTEKDTQEGI ITVKKPLDYE SRRLYTLKVE AENTHVDPRF YYLGPFKDTT IVKISIEDVD EPPVFSRSSY LFEVH Antigen sequence identity: Mouse 97%; Rat 97%.
CDH10 is a type II classical cadherin from the cadherin superfamily, integral membrane proteins that mediate calcium-dependent cell-cell adhesion. Mature cadherin proteins are composed of a large N-terminal extracellular domain, a single membrane-spanning domain, and a small, highly conserved C-terminal cytoplasmic domain. The extracellular domain consists of 5 subdomains, each containing a cadherin motif, and appears to determine the specificity of the protein's homophilic cell adhesion activity. Type II (atypical) cadherins are defined based on their lack of a HAV cell adhesion recognition sequence specific to type I cadherins. This particular cadherin is predominantly expressed in brain and is putatively involved in synaptic adhesions, axon outgrowth and guidance.
Specifications
CDH10 | |
Polyclonal | |
Unconjugated | |
Cdh10 | |
A830016G23Rik; C030003B10Rik; C030011H18Rik; Cadherin; cadherin 10; cadherin 10 type 2; cadherin 10, type 2 (T2-cadherin); cadherin-10; Cdh10; T2-cadherin | |
Rabbit | |
Affinity chromatography | |
RUO | |
1008 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y6N8 | |
Cdh10 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 250-394. Recombinant protein control fragment (Product #RP-109975). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction