Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDH16 Mouse anti-Human, Clone: SPM594, Abnova™

Catalog No. 89933915 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-915 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-915 Supplier Abnova Corporation Supplier No. MAB13054
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full length recombinant human CDH16.

This gene is a member of the cadherin superfamily, genes encoding calcium-dependent, membrane-associated glycoproteins. Mapped to a previously identified cluster of cadherin genes on chromosome 16q22.1, the gene localizes with superfamily members CDH1, CDH3, CDH5, CDH8 and CDH11. The protein consists of an extracellular domain containing 6 cadherin domains, a transmembrane region and a truncated cytoplasmic domain but lacks the prosequence and tripeptide HAV adhesion recognition sequence typical of most classical cadherins. Expression is exclusively in kidney, where the protein functions as the principal mediator of homotypic cellular recognition, playing a role in the morphogenic direction of tissue development. [provided by RefSeq]

Specifications

Antigen CDH16
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone SPM594
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant human CDH16.
Dilution Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS (0.05% BSA, 0.05% sodium azide).
Gene Symbols CDH16
Host Species Mouse
Immunogen Recombinant protein corresponding to full length of human CDH16.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1014
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.