Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDH16 Polyclonal Antibody

Rabbit Polyclonal Antibody

$209.50 - $511.50

Specifications

Antigen CDH16
Concentration 1 mg/mL
Content And Storage -20°C
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIPA5118987
View Documents
Invitrogen™
PA5118987
200 μL
Each for $511.50
Only null left
Add to Cart
 
PIPA5122106
View Documents
Invitrogen™
PA5122106
20 μL
Each for $209.50
Only null left
Add to Cart
 
Description

Description

CDH16 Polyclonal Antibody for Western Blot, IHC (P)

The cadherins are a family of Ca2+-dependent adhesion molecules that function to mediate cell-cell binding critical to the maintenance of structure and morphogenesis. Cadherins each contain a large extracellular domain at the N-terminus, which is characterized by a series of five homologous repeats, the most distal of which is thought to be responsible for binding specificity. Cadherin-16, also known as Ksp-Cadherin, is a type I membrane protein which is kidney-specific. Cadherin-16 is expressed exclusively in the basolateral membrane of renal tubular epithelial cells. The human Ksp-cadherin gene (Cadherin-16) maps to chromosome 16q21-proximal 16q22. The mouse Ksp-cadherin gene was localized to a highly syntenic region of distal Chromosome 8.
Specifications

Specifications

CDH16
-20°C
Polyclonal
Liquid
IgG
Mouse
CDH16
Cadherin; cadherin 16; cadherin 16, KSP-cadherin; cadherin-16; Cadherin-16 (CDH16); CDH 16; CDH16; Kidney-specific cadherin; KSP-cadherin; Ksp-cadherin antibody; UNQ695/PRO1340
CDH16
Primary
Antigen affinity chromatography
1 mg/mL
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.05% ProClin 300
O88338
12556
Recombinant protein Cadherin 16. The antigen corresponds to amino acid range 353-617 of the target protein.
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.