Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDH17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554843
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554843 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554843 Supplier Invitrogen™ Supplier No. PA554843
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: INNVMYFQIN NKTGAISLTR EGSQELNPAK NPSYNLVISV KDMGGQSENS FSDTTSVDII VTENIWKAPK PVEMVENSTD P Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.

CDH17 gene is a member of the cadherin superfamily, genes encoding calcium-dependent, membrane-associated glycoproteins. The encoded protein is cadherin-like, consisting of an extracellular region, containing 7 cadherin domains, and a transmembrane region but lacking the conserved cytoplasmic domain. The protein is a component of the gastrointestinal tract and pancreatic ducts, acting as an intestinal proton-dependent peptide transporter in the first step in oral absorption of many medically important peptide-based drugs. The protein may also play a role in the morphological organization of liver and intestine. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDH17
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cdh17
Gene Accession No. Q12864
Gene Alias BILL-cadherin; Cadherin; cadherin 17; cadherin 17, LI cadherin (liver-intestine); cadherin-16; cadherin-17; CDH16; CDH17; FLJ26931; HPT1; HPT-1; HPT-1 cadherin; HPT-1/LI; human intestinal peptide-associated transporter HPT-1; human peptide transporter 1; intestinal peptide-associated transporter HPT-1; LI cadherin; LI-cadherin; Liver-intestine cadherin; MGC138218; MGC142024; P130
Gene Symbols Cdh17
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CDH17. Recombinant protein control fragment (Product #RP-93741).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1015
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.