Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDH19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563288
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563288 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563288 Supplier Invitrogen™ Supplier No. PA563288
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DYSIEEDDSQ TFDIITNHET QEGIVILKKK VDFEHQNHYG IRAKVKNHHV PEQLMKYHTE ASTTFIKIQV Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 74%.

This gene is one of three related type II cadherin genes situated in a cluster on chromosome 18. The encoded protein is a calcium dependent cell-cell adhesion glycoprotein containing five extracellular cadherin repeats. Loss of cadherins may be associated with cancer formation. Alternative splicing results in multiple transcript variants for this gene. [provided by RefSeq, Aug 2012].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDH19
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDH19
Gene Accession No. Q9H159
Gene Alias Cadherin; cadherin 19; cadherin 19, type 2; cadherin-19; CDH19; CDH7; CDH7L2; UNQ478/PRO941
Gene Symbols CDH19
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CDH19. Recombinant protein control fragment (Product #RP-101114).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28513
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.