Learn More
CDH2 Mouse anti-Canine, Human, Mouse, Clone: OTI2G7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503933
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CDH2 encodes a classical cadherin and member of the cadherin superfamily. Alternative splicing results in multiple transcript variants, at least one of which encodes a preproprotein is proteolytically processed to generate a calcium-dependent cell adhesion molecule and glycoprotein. This protein plays a role in the establishment of left-right asymmetry, development of the nervous system and the formation of cartilage and bone.Specifications
| CDH2 | |
| Monoclonal | |
| Unconjugated | |
| CDH2 | |
| CD325, CDHN, CDw325, NCAD | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1000, 12558, 480169 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2G7 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P15116, P19022 | |
| CDH2 | |
| Full length human recombit protein of human CDH2 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Canine, Human, Mouse | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.