Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDH2 Mouse anti-Human, Unconjugated, Clone: 8C11, Abnova™

Catalog No. 89337352 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-352 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-352 Supplier Abnova Corporation Supplier No. MAB20855
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

CDH2 Monoclonal antibody specifically detects CDH2 in Human samples. It is validated for WB, IHC-P, IF, Flow Cyt.

Specifications

Antigen CDH2
Applications Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone 8C11
Conjugate Unconjugated
Description cadherin 2, type 1, N-cadherin (neuronal)
Gene CDH2
Gene Alias CD325, CDHN, CDw325, NCAD
Host Species Mouse
Immunogen Recombinant protein corresponding to extracellular domain of human CDH2
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 1000
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.