Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CDH20 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553309
Description
Immunogen sequence: PFQDTTTVHI SVEDVDEPPV FEPGFYFVEV PEDVAIGTTI QIISAKDPDV TNNSIRYSID RSSDPGRFFY VDITTGALMT ARPLDREEFS WHNITVLAME MNNPSQVGSV PVTIKV Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
This gene is a type II classical cadherin from the cadherin superfamily and one of three cadherin 7-like genes located in a cluster on chromosome 18. The encoded membrane protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Type II (atypical) cadherins are defined based on their lack of a HAV cell adhesion recognition sequence specific to type I cadherins. Since disturbance of intracellular adhesion is a prerequisite for invasion and metastasis of tumor cells, cadherins are considered prime candidates for tumor suppressor genes.
Specifications
| CDH20 | |
| Polyclonal | |
| Unconjugated | |
| CDH20 | |
| Cad20; Cadherin; cadherin 20; cadherin 20, type 2; cadherin 7; cadherin-20; Cadherin-7; Cdh20; Cdh7; CDH7L3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 28316 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HBT6 | |
| CDH20 | |
| Recombinant protein corresponding to Human CDH20. Recombinant protein control fragment (Product #RP-90036). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction