Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CDH20 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575933
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene is a type II classical cadherin from the cadherin superfamily and one of three cadherin 7-like genes located in a cluster on chromosome 18. The encoded membrane protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Type II (atypical) cadherins are defined based on their lack of a HAV cell adhesion recognition sequence specific to type I cadherins. Since disturbance of intracellular adhesion is a prerequisite for invasion and metastasis of tumor cells, cadherins are considered prime candidates for tumor suppressor genes.
Specifications
CDH20 | |
Polyclonal | |
Unconjugated | |
CDH20 | |
Cad20; Cadherin; cadherin 20; cadherin 20, type 2; cadherin 7; cadherin-20; Cadherin-7; Cdh20; Cdh7; CDH7L3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23836, 28316, 363948 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5DWV1, Q9HBT6, Q9Z0M3 | |
CDH20 | |
Synthetic peptide corresponding to amino acids 97-142 of Human CDH20. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction