Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDH22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552919
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552919 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552919 Supplier Invitrogen™ Supplier No. PA552919
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MYQFSIQESA PIGTAVGRVK AEDSDVGENT DMTYHLKDES SSGGDVFKVT TDSDTQEAII VVQKRLDFES QPVHTVILEA LNKFVDPRFA DLGTFRDQAI VRVAVTDVDE P Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

This gene is a member of the cadherin superfamily. The gene product is composed of five cadherin repeat domains and a cytoplasmic tail similar to the highly conserved cytoplasmic region of classical cadherins. Expressed predominantly in the brain, this putative calcium-dependent cell adhesion protein may play an important role in morphogenesis and tissue formation in neural and non-neural cells during development and maintenance of the brain and neuroendocrine organs. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDH22
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDH22
Gene Accession No. Q9UJ99
Gene Alias C20orf25; Cadherin; cadherin 22; cadherin 22, type 2; cadherin-22; cadherin-like 22; CDH22; dJ998H6.1; ortholog of rat PB-cadherin; PB-cadherin; Pituitary and brain cadherin
Gene Symbols CDH22
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CDH22. Recombinant protein control fragment (Product #RP-90328).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64405
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.