Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CDH22 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569617
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 91%.
This gene is a member of the cadherin superfamily. The gene product is composed of five cadherin repeat domains and a cytoplasmic tail similar to the highly conserved cytoplasmic region of classical cadherins. Expressed predominantly in the brain, this putative calcium-dependent cell adhesion protein may play an important role in morphogenesis and tissue formation in neural and non-neural cells during development and maintenance of the brain and neuroendocrine organs. [provided by RefSeq, Jul 2008].
Specifications
CDH22 | |
Polyclonal | |
Unconjugated | |
CDH22 | |
C20orf25; Cadherin; cadherin 22; cadherin 22, type 2; cadherin-22; cadherin-like 22; CDH22; dJ998H6.1; ortholog of rat PB-cadherin; PB-cadherin; Pituitary and brain cadherin | |
Rabbit | |
Affinity Chromatography | |
RUO | |
64405 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UJ99 | |
CDH22 | |
synthetic peptide directed towards the middle region of human CDH22 (aa 111-160). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction