Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDH23 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543398
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543398 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543398 Supplier Invitrogen™ Supplier No. PA543398
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DHQGVITRKV NIQVGDVNDN APTFHNQPYS VRIPENTPVG TPIFIVNATD Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

This gene is a member of the cadherin superfamily, whose genes encode calcium dependent cell-cell adhesion glycoproteins. The encoded protein is thought to be involved in stereocilia organization and hair bundle formation. The gene is located in a region containing the human deafness loci DFNB12 and USH1D. Usher syndrome 1D and nonsyndromic autosomal recessive deafness DFNB12 are caused by allelic mutations of this cadherin-like gene. Alternative splice variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDH23
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CDH23
Gene Accession No. Q9H251
Gene Alias 4930542A03Rik; age related hearing loss 1; ahl; ahl1; bob; bobby; bus; bustling; Cadherin; cadherin 23; cadherin 23 (otocadherin); cadherin related 23; cadherin-23; cadherin-like 23; cadherin-related 23; cadherin-related family member 23; Cdh23; CDHR23; DFNB12; DKFZp434P2350; FLJ00233; FLJ36499; KIAA1774; KIAA1812; mdfw; MGC102761; modifier of deaf waddler; nmf112; nmf181; nmf252; otocadherin; OTTHUMP00000044780; sals; UNQ1894/PRO4340; USH1D; USH1H; v; W; waltzer; Waltzing
Gene Symbols CDH23
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human CDH23 (aa 109-158).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64072
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.