Learn More
Invitrogen™ CDH24 Polyclonal Antibody

Description
Immunogen sequence: TVLATELGWS WGPERGWVPL LVAEWSAPAA PPQRSPVGSA VGIPQDSSAQ AS Highest antigen sequence indentity to the following orthologs: Rat - 31%, Mouse - 31%.
Specifications
Specifications
| Antigen | CDH24 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CDH24 |
| Gene Accession No. | Q86UP0 |
| Gene Alias | 1700040A22Rik; Cadherin; cadherin 24; cadherin 24, type 2; cadherin-24; cadherin-like 24; CDH11L; CDH24; ENSMUSG00000022188; EY-cadherin; UNQ2834/PRO34009 |
| Gene Symbols | CDH24 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.