Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDH24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145076
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145076 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145076 Supplier Invitrogen™ Supplier No. PA5145076
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TVLATELGWS WGPERGWVPL LVAEWSAPAA PPQRSPVGSA VGIPQDSSAQ AS Antigen sequence identity: Mouse 31%; Rat 31%.

Cadherins are calcium-dependent cell adhesion proteins. They preferentially interact with themselves in a homophilic manner in connecting cells; cadherins may thus contribute to the sorting of heterogeneous cell types. Cadherin-24 mediate strong cell-cell adhesion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDH24
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDH24
Gene Accession No. Q86UP0
Gene Alias 1700040A22Rik; Cadherin; cadherin 24; cadherin 24, type 2; cadherin-24; cadherin-like 24; CDH11L; CDH24; ENSMUSG00000022188; EY-cadherin; UNQ2834/PRO34009
Gene Symbols CDH24
Host Species Rabbit
Immunogen Recombinant protein fragment of Human CDH24 (Product #RP-106510)
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64403
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.