Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDH24 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA571647
Description
Cadherins are calcium-dependent cell adhesion proteins. They preferentially interact with themselves in a homophilic manner in connecting cells; cadherins may thus contribute to the sorting of heterogeneous cell types. Cadherin-24 mediate strong cell-cell adhesion.
Specifications
| CDH24 | |
| Polyclonal | |
| Unconjugated | |
| CDH24 | |
| Cadherin-24; CDH24; CDH11L | |
| Rabbit | |
| Protein A | |
| RUO | |
| 64403 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.46 mg/mL | |
| PBS with 0.09% sodium azide | |
| Q86UP0 | |
| CDH24 | |
| KLH conjugated synthetic peptide between 54-81 amino acids from the N-terminal region of human CDH24 | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction