Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDH26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103396
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103396 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103396 Supplier Thermo Scientific Supplier No. PA5103396
Add to Cart
Add to Cart

CDH26 Polyclonal Antibody

CDH26 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen CDH26
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CDH26
Gene Accession No. P59862, Q8IXH8
Gene Alias Cadherin; cadherin 26; cadherin-like 26; cadherin-like protein 26; Cadherin-like protein VR20; CDH26; Gm1010; VR20
Gene Symbols CDH26
Host Species Rabbit
Immunogen A synthesized peptide derived from human CDH26(Accession Q8IXH8), corresponding to amino acid residues L15-E65.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 311693, 381409, 60437
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.