Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDH26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553372
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553372 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553372 Supplier Invitrogen™ Supplier No. PA553372
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VIIIHAVDDG FPPQTATGTL MLFLSDINDN VPTLRPRSRY MEVCESAVHE PLHIEAEDPD LEPFSDPFTF ELDNTWGNAE DTWKLGRNWG QSVELLTLRS LPRGNYLVPL FIGDKQGLSQ KQTVHVRICP CASGLTCVEL ADAEVGL Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 56%.

Cadherins are a family of adhesion molecules that mediate Ca2+-dependent cell-cell adhesion in all solid tissues and modulate a wide variety of processes, including cell polarization and migration. Cadherin domains occur as repeats in the extracellular region and are thought to contribute to the sorting of heterogeneous cell types and the maintenance of orderly structures such as epithelium. This gene encodes a cadherin domain-containing protein whose specific function has not yet been determined. Alternative splicing occurs at this locus and two transcript variants, encoding distinct proteins, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDH26
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDH26
Gene Accession No. Q8IXH8
Gene Alias Cadherin; cadherin 26; cadherin-like 26; cadherin-like protein 26; Cadherin-like protein VR20; CDH26; Gm1010; VR20
Gene Symbols CDH26
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CDH26. Recombinant protein control fragment (Product #RP-90175).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60437
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.