Learn More
CDH4 Mouse anti-Human, Clone: OTI1B7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF804801

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CDH4 is a classical cadherin from the cadherin superfamily. It is a calcium-dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Based on studies in chicken and mouse, this cadherin is thought to play an important role during brain segmentation and neuronal outgrowth. In addition, a role in kidney and muscle development is indicated. Of particular interest are studies showing stable cis-heterodimers of cadherins 2 and 4 in cotransfected cell lines. Previously thought to interact in an exclusively homophilic manner, this is the first evidence of cadherin heterodimerization.Specifications
CDH4 | |
Monoclonal | |
Unconjugated | |
CDH4 | |
CAD4, R-CAD, RCAD | |
Mouse | |
Affinity Chromatography | |
RUO | |
1002 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1B7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P55283 | |
CDH4 | |
Human recombit protein fragment corresponding to amino acids 170-428 of human CDH4 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.