Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ cdhr1a Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5144076
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5144076 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5144076 Supplier Invitrogen™ Supplier No. PA5144076
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

cdhr1a Polyclonal Antibody for IHC (P)

This gene is a member of the cadherin superfamily of calcium-dependent cell-cell adhesion molecules. This non-classical cadherin appears to be exclusively expressed in the mitral and tufted cells in the main and accessory olfactory bulbs of the brain, suggesting a possible role in the formation and maintenance of neuronal networks. Alternatively spliced transcript variants encoding different isoform have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen cdhr1a
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene cdhr1a
Gene Accession No. Q6B3P0
Gene Alias Cadherin; cadherin related family member 1; cadherin-related family member 1; cadherin-related family member 1a; cdhr1; cdhr1a; CORD15; DKFZp434A132; KIAA1775; mKIAA1775; MT-protocadherin; pcdh21; Photoreceptor cadherin; photoreceptor protocadherin; prCAD; protocadherin 21; protocadherin; prCAD; protocadherin-21; RP65
Gene Symbols cdhr1a
Host Species Rabbit
Immunogen Zebrafish CDHR1 between 26-247.
Purification Method Antigen affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 449008
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.