Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDHR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144695
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144695 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144695 Supplier Invitrogen™ Supplier No. PA5144695
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ATVPVTIRIV DLNNHPPTFY GESGPQNRFE LSMNEHPPQG EILRGLKITV NDSDQGANAK FNLQLVGPRG IFRVVPQTVL NEAQVTIIVE NSAAIDFEKS KVLTFKLLAV EVNTPEKFSS TADV Antigen sequence identity: Mouse 94%; Rat 93%.

This gene is a member of the cadherin superfamily of calcium-dependent cell-cell adhesion molecules. This non-classical cadherin appears to be exclusively expressed in the mitral and tufted cells in the main and accessory olfactory bulbs of the brain, suggesting a possible role in the formation and maintenance of neuronal networks. Alternatively spliced transcript variants encoding different isoform have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDHR1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDHR1
Gene Accession No. Q96JP9
Gene Alias Cadherin; cadherin related family member 1; cadherin-related family member 1; cadherin-related family member 1a; cdhr1; cdhr1a; CORD15; DKFZp434A132; KIAA1775; mKIAA1775; MT-protocadherin; pcdh21; Photoreceptor cadherin; photoreceptor protocadherin; prCAD; protocadherin 21; protocadherin; prCAD; protocadherin-21; RP65
Gene Symbols CDHR1
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 335-458. Recombinant protein control fragment (Product #RP-109725).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92211
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.