Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDHR3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578657
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578657 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578657 Supplier Invitrogen™ Supplier No. PA578657
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: transfected CDHR3 in HEK293FT Predicted Reactivity: Mouse (80%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Cadherins are calcium dependent cell adhesion proteins. They preferentially interact with themselves in a homophilic manner in connecting cells; cadherins may thus contribute to the sorting of heterogeneous cell type.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDHR3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene CDHR3
Gene Accession No. Q6ZTQ4
Gene Alias Cadherin; cadherin related family member 3; cadherin-like protein 28; Cadherin-related family member 3; CDH28; CDHR3; FLJ23834; FLJ43271; MGC133292; MGC133293
Gene Symbols CDHR3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the Extracellular domain of human FLJ23834. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222256
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.