Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDHR4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584996
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584996 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584996 Supplier Invitrogen™ Supplier No. PA584996
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ATLDYKLWFR SSSNPASLCL YDRVLEVNAT LDCDTPGACF QHAASILVLD GGQPQMTTEV PVLVMVTPIN EFSPACAPRT FRVQE Highest antigen sequence indentity to the following orthologs: Rat - 27%, Mouse - 80%.

Cadherins are calcium-dependent cell adhesion proteins. They preferentially interact with themselves in a homophilic manner in connecting cells; cadherins may thus contribute to the sorting of heterogeneous cell types .
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDHR4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDHR4
Gene Accession No. A6H8M9
Gene Alias Cadherin; cadherin related family member 4; cadherin-like 29; cadherin-like protein 29; Cadherin-like protein UNQ9392/PRO34300; cadherin-related family member 4; CDH29; CDHR4; PRO34300; UNQ9392/PRO34300; VLLR9392
Gene Symbols CDHR4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CDHR4. Recombinant protein control fragment (Product #RP-108280).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389118
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.