Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDHR5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547590
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547590 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547590 Supplier Invitrogen™ Supplier No. PA547590
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human MUCDHL is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

This gene is a novel mucin-like gene that is a member of the cadherin superfamily. While encoding nonpolymorphic tandem repeats rich in proline, serine and threonine similar to mucin proteins, the gene also contains sequence encoding calcium-binding motifs found in all cadherins. The role of the hybrid extracellular region and the specific function of this protein have not yet been determined. Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDHR5
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene CDHR5
Gene Accession No. Q8VHF2
Gene Alias 1810074H01Rik; AI481143; Cadherin; cadherin related family member 5; cadherin-related family member 5; CDHR5; differentiation-associated catenin regulator; GP100; MLPCDH; MUCDHL; mucin and cadherin like; mucin and cadherin-like protein; Mucin-like protocadherin; Mupcdh; MU-PCDH; mu-protocadherin; UNQ2781/PRO7168
Gene Symbols CDHR5
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse MUCDHL Gln29-Ala479.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 72040
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.