Learn More
Invitrogen™ CDHR5 Polyclonal Antibody

Description
Immunogen sequence: CIQAQYHGAV PTGHILPSPL VLRPGPIYAE DGDRGINQPI IYSIFRGNVN GTFIIHPDSG NLTVARSVPS PMTFLLLVKG QQADLARYSV TQVTVEAVAA AGSPPRFPQS LYRGTVARGA GAGVVVKDA Highest antigen sequence indentity to the following orthologs: Mouse - 60%, Rat - 65%.
Specifications
Specifications
| Antigen | CDHR5 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CDHR5 |
| Gene Accession No. | Q9HBB8 |
| Gene Alias | 1810074H01Rik; AI481143; Cadherin; cadherin related family member 5; cadherin-related family member 5; CDHR5; differentiation-associated catenin regulator; GP100; MLPCDH; MUCDHL; mucin and cadherin like; mucin and cadherin-like protein; Mucin-like protocadherin; Mupcdh; MU-PCDH; mu-protocadherin; UNQ2781/PRO7168 |
| Gene Symbols | CDHR5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.