Learn More
Invitrogen™ CDHR5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589483
Description
Immunogen sequence: QAQYCSVNKD IFEVEENTNV TEPLVDIHVP EGQEVTLGAL STPFAFRIQG NQLFLNVTPD YEEKSLLEAQ LLCQSGGTLV TQLRVFVSVL DVNDNAPEFP FKTKEIRVEE DTKVNSTVIP ETQLQAEDRD KDDILFYTLQ EMTAGASDYF SLVSVNRPAL RLDRPLDFYE RPNMTFWLLV RDTPGENVEP SHTAT; Positive Samples: Mouse kidney, Rat kidney; Cellular Location: Cell membrane, Single-pass type I membrane protein.
This gene is a novel mucin-like gene that is a member of the cadherin superfamily. While encoding nonpolymorphic tandem repeats rich in proline, serine and threonine similar to mucin proteins, the gene also contains sequence encoding calcium-binding motifs found in all cadherins. The role of the hybrid extracellular region and the specific function of this protein have not yet been determined. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
CDHR5 | |
Polyclonal | |
Unconjugated | |
CDHR5 | |
1810074H01Rik; AI481143; Cadherin; cadherin related family member 5; cadherin-related family member 5; CDHR5; differentiation-associated catenin regulator; GP100; MLPCDH; MUCDHL; mucin and cadherin like; mucin and cadherin-like protein; Mucin-like protocadherin; Mupcdh; MU-PCDH; mu-protocadherin; UNQ2781/PRO7168 | |
Rabbit | |
Affinity chromatography | |
RUO | |
171554, 72040 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.9 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8VHF2, Q9JIK1 | |
CDHR5 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 26-220 of human CDHR5 (NP_1125542). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.