Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDIP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561940
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561940 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561940 Supplier Invitrogen™ Supplier No. PA561940
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CLINDFKDVT HTCPSCKAYI YTYKLLC Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.

The p53 tumor-suppressor gene integrates numerous signals that control cell life and death; loss of its functions contributes to the development of most cancers. CDIP is a novel pro-apoptotic target gene whose inhibition abrogates p53-mediated apoptotic responses. Overexpression of CDIP induced apoptosis in transfected cells while siRNA suppression of caspase-8 mRNA blocked this CDIP-induced apoptosis, indicating that the CDIP-dependent apoptosis pathway proceeds through extrinsic cell death pathway. CDIP may thus represent a novel target for drug design to maximize p53 response and sensitize tumor cells to cancer therapy. Multiple isoforms of CDIP are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDIP1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDIP1
Gene Accession No. Q9H305
Gene Alias 2700048O17Rik; 5730403B10Rik; AU018521; AW742692; C16orf5; CDIP; CDIP1; cell death inducing protein; cell death inducing Trp53 target 1; cell death involved p53-target; cell death-inducing p53 target 1; cell death-inducing p53-target protein 1; Cell death-inducing protein; I1; Lipopolysaccharide-induced tumor necrosis factor-alpha-like protein; LITAFL; LITAF-like protein; Transmembrane protein I1
Gene Symbols CDIP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CDIP1. Recombinant protein control fragment (Product #RP-101138).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29965
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.