Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdk-associated cullin1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980392
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0392 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0392 Supplier Bioss Supplier No. BS0614R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cell cycle associated protein capable of promoting cell proliferation through the activation of CDK2 at the G1/S phase transition.

Specifications

Antigen Cdk-associated cullin1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CACUL1
Gene Accession No. Q86Y37
Gene Alias 2700078E11Rik; 2810417M16Rik; 9830127L17Rik; AI450346; C10orf46; CAC1; Cacul1; CDK2 associated cullin domain 1; CDK2 associated, cullin domain 1; CDK2-associated and cullin domain-containing protein 1; CDK2-associated, cullin domain 1; Cdk-associated cullin1; cullin; D130033C15Rik; RGD1308127
Gene Symbols CACUL1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Cdk-associated cullin1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143384
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.