Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDK1/2/3, p Thr14 Antibody (SR1566), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | CDK1/2/3 |
---|---|
Clone | SR1566 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Description
CDK1/2/3 Monoclonal antibody specifically detects CDK1/2/3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
CDK1/2/3 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
IgG | |
Affinity purified |
SR1566 | |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
A synthesized peptide derived from human Phospho-CDK1/2/3 [Thr14] (Uniprot #: P06493, P24941, Q00526) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title