Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDK1/CDK2 Monoclonal Antibody (AN21.2)
GREENER_CHOICE

Catalog No. PIMA191380
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA191380 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA191380 Supplier Invitrogen™ Supplier No. MA191380
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Recommended positive controls: HeLa nuclear extract.

Voltage dependent anion channel required for acidification and functions of the Golgi apparatus that may function in counter-ion conductance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDK1/CDK2
Applications ELISA, Western Blot
Classification Monoclonal
Clone AN21.2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CDK1
Gene Accession No. P06493, P24941
Gene Alias A630093N05Rik; Cdc2; CDC28A; Cdc2a; cdc2-related protein kinase; Cdk1; CDK2; CDKN1; CDKN2; cell cycle controller CDC2; cell cycle p34 CDC2 kinase protein; cell division control protein 2 homolog; cell division cycle 2 homolog A; cell division cycle 2, G1 to S and G2 to M; Cell division cycle control protein 2; cell division cycle control protein 2a; cell division protein kinase 1; Cell division protein kinase 2; cyclin dependent kinase 1; cyclin dependent kinase 2; cyclin dependent kinase 2-alpha; cyclin-dependent kinase 1; Cyclin-dependent kinase 2; p33 protein kinase; p33(CDK2); p34 protein kinase; p34>CDC2>; P34CDC2
Gene Symbols CDK1, CDK2
Host Species Mouse
Immunogen Human cdk2.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1017, 983
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.