Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Cdk1/p34 cdc2 Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and kinase assay applications.

Supplier:  Epredia™ MS110R1

Catalog No. MS110R1


Description

Description

p34cdc2(a catalytic subunit of Maturation Promoting Factor) plays a crucial role during cell division and is most active during mitosis. It is a serine/threonine kinase, which is activated by cyclin through dephosphorylation of tyrosine residues. p34cdc2 is inactivated by a tyrosine kinase.

Host Species: Mouse

Clone: A17.1.1

Isotype: IgG2a

Species Reactivity: Human, Mouse, Rat, Guinea Pig, Woodchuck, Xenopus and Chicken.

Epitope: aa 220-227

Immunogen: C-Terminal 2/3 of Xenopus CDC2 expressed in E. coli

Molecular Weight: 34 kDa

Positive Control: HeLa, MAD109, PC12 or NIH-3T3 cells; Tonsil or lymph node

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Kinase Assay
  • Inhibits activation of p34cdc2 kinase by cyclins

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Cdk1/p34superscriptcdc2superscript Ab-1
Monoclonal
Unconjugated
Mouse
1 mL
Cell Cycle
Chicken, Guinea Pig, Human, Mouse, Rat, Woodchuck, Xenopus species
IgG2a
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
A17.1.1
Ready-to-use for IHC (UVLP)
C-Terminal 2/3 of Xenopus CDC2 expressed in E. coli
RUO
Primary
Prediluted
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.