Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDK1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA596129
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596129 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596129 Supplier Thermo Scientific Supplier No. PA596129

Rabbit Polyclonal Antibody

Positive Samples: HL-60, HeLa, THP-1, Rat spleen; Cellular Location: Cytoplasm, Mitochondrion, Nucleus, centrosome, cytoskeleton, microtubule organizing center, spindle Immunogen sequence: ATKKPLFHGD SEIDQLFRIF RALGTPNNEV WPEVESLQDY KNTFPKWKPG SLASHVKNLD ENGLDLLSKM LIYDPAKRIS GKMALNHPYF NDLDNQIKKM

CDK1 or Cell Division Control protein 1 is essential for the completion of START, the controlling event in the cell cycle that is required to initiate mitosis. CDK1 is a catalytic subunit of a protein kinase complex, called the M-Phase Promoting Factor that induces entry into mitosis and is universal among eukaryotes. Phosphorylation of Bcl-2 in G2/M phase-arrested cells following photodynamic therapy with hypericin involves a CDK1-mediated signal and delays the onset of apoptosis. Therapeutic potential of CDK inhibitor NU2058 in androgen-independent prostate cancer has also been demonstrated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDK1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide, pH 7.3
Gene CDK1
Gene Accession No. P06493, P11440, P39951
Gene Alias 5363, ABL1, cdc, CDC2, cdc-2, cdc2 kinase, CDC28A, Cdc2a, cdc2-a, cdc2Dm, cdc2x1.1, CDCDm, CDK1, Cdk-1, cdk1.S, CDK1/CDC2, CDK1-A, cdk1-b, Cdk1-PA, CDKN1, cell cycle controller CDC2, cell cycle p34 CDC2 kinase protein, cell division control, cell division control protein 2 homolog, Cell division control protein 2 homolog 1, cell division control protein 2-A, cell division cycle 2 homolog A, cell division cycle 2 like, cell division cycle 2, G1 to S and G2 to M, Cell division cycle control protein 2, cell division cycle control protein 2a, cell division protein kinase 1, Cell division protein kinase 1-A, CG5363, CG5363-PA, cyclin dependent kinase, cyclin dependent kinase 1, cyclin-dependent kinase, Cyclin-dependent kinase 1, cyclin-dependent kinase 1 S homeolog, cyclin-dependent kinase 1-A, D2366, Dcdc2, dCdk1, DKFZp686L20222, Dm cdc2, Dmcdc2, DmCdk1, Dmel\CG5363, Dmel_CG5363, DNA polymerase delta catalytic subunit, DNA polymerase III, DNA-directed DNA polymerase delta POL3, group 4, HPR6, KCIP-1, l(2)31Eh, LD38718p, maturation-promoting factor, MGC111195, MGC53829, MPF, OTTHUMP00000019660, P34 CDK1, P34 MGC111195, p34 protein kinase, p34 protein kinase 1, p34<CDC2>, p34cdc2, p34-cdc2 homologue, POL3, Protein 1054, protein 2, PSTAIR, TEX1, unnamed protein product, xcdc2, XELAEV_18036802mg, YDL102W
Gene Symbols CDK1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 198-297 of human CDK1 (NP_0017771)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12534, 54237, 983
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.