Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDK10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142464
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142464 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142464 Supplier Thermo Scientific Supplier No. PA5142464
Add to Cart
Add to Cart
This item is not returnable. View return policy

CDK10 Polyclonal Antibody

CDK10 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen CDK10
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene CDK10
Gene Accession No. Q15131
Gene Alias BC017131; CDC2-related protein kinase; Cdk10; Cell division protein kinase 10; cyclin dependent kinase 10; cyclin-dependent kinase (CDC2-like) 10; cyclin-dependent kinase 10; cyclin-dependent kinase related protein; PISSLRE; serine/threonine protein kinase PISSLRE; serine/threonine-protein kinase PISSLRE
Gene Symbols CDK10
Host Species Goat
Immunogen Peptide with sequence AEPDLECEQIRLK (aa 2-14).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8558
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.