Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CDK10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101169
Description
Antibody detects endogenous levels of total CDK10.
The protein encoded by this gene belongs to the CDK subfamily of the Ser/Thr protein kinase family. The CDK subfamily members are highly similar to the gene products of S. cerevisiae cdc28, and S. pombe cdc2, and are known to be essential for cell cycle progression. This kinase has been shown to play a role in cellular proliferation and its function is limited to cell cycle G2-M phase. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
CDK10 | |
Polyclonal | |
Unconjugated | |
CDK10 | |
BC017131; CDC2-related protein kinase; Cdk10; Cell division protein kinase 10; cyclin dependent kinase 10; cyclin-dependent kinase (CDC2-like) 10; cyclin-dependent kinase 10; cyclin-dependent kinase related protein; PISSLRE; serine/threonine protein kinase PISSLRE; serine/threonine-protein kinase PISSLRE | |
Rabbit | |
Affinity chromatography | |
RUO | |
234854, 361434, 8558 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q15131, Q3UMM4, Q4KM47 | |
CDK10 | |
A synthesized peptide derived from human CDK10(Accession Q15131), corresponding to amino acid residues N43-L93. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction