Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDK11A/CDK11B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104521
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104521 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104521 Supplier Invitrogen™ Supplier No. PA5104521
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CDK11.

CDK11 is a member of the cyclin-dependent kinase 11 family which is known to be essential for eukaryotic cell cycle control and it is in close proximity to CDC2L2, a nearly identical gene in the same chromosomal region. CDK11 is a negative regulator of normal cell cycle progression and Its expression is regulated during murine embryogenesis, and its activity is coordinately regulated with that of p34 (cdc2) during the cell cycle. CDK11 gene may harbor 1 or more tumor suppressor genes that affected by chromosome 1p36 modifications in neuroblastoma. CDK11 could be cleaved by caspases and was demonstrated to play roles in cell apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDK11A/CDK11B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CDK11B
Gene Accession No. P21127, P24788, Q9UQ88
Gene Alias AA989746; Cdc11b; Cdc2l1; CDC2L2; CDC2L3; CDC-related protein kinase p58; CDK11; CDK11A; Cdk11b; CDK11-p110; CDK11-p46; CDK11-p58; cell division cycle 2 homolog-like 1; cell division cycle 2 homolog-like 2; cell division cycle 2-like 1; cell division cycle 2-like 1 (PITSLRE proteins); cell division cycle 2-like 2; cell division cycle 2-like 2 (PITSLRE proteins); cell division cycle 2-like protein kinase 1; Cell division cycle 2-like protein kinase 2; cell division protein kinase 11; Cell division protein kinase 11A; cell division protein kinase 11B; CLK-1; cyclin dependent kinase 11A; cyclin dependent kinase 11B; cyclin-dependent kinase 11; Cyclin-dependent kinase 11A; cyclin-dependent kinase 11B; Galactosyltransferase-associated protein kinase p58/GTA; p58; p58 CLK-1; p58CDC2L1; p58CLK-1; p58GTA; PITSLRE; PITSLRE B; PITSLRE protein kinase beta; PITSLRE proteins; PITSLRE serine/threonine-protein kinase CDC2L1; PITSLRE serine/threonine-protein kinase CDC2L2; PITSLREA; PITSLREB; PK58
Gene Symbols CDK11A, CDK11B
Host Species Rabbit
Immunogen A synthesized peptide derived from human CDK11A(Accession Q9UQ88), corresponding to amino acid residues D212-D262.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12537, 728642, 984
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.