Learn More
Invitrogen™ CDK11B Polyclonal Antibody

Description
This antibody detects endogenous protein at a molecular weight of 110 and 92 kDa. Purity is >95% by SDS-PAGE.
Specifications
Specifications
| Antigen | CDK11B |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide; pH 7.2 |
| Gene | CDK11B |
| Gene Accession No. | P21127, P24788, P46892 |
| Gene Alias | AA989746; Cdc11b; CDC2L1; Cdc2l2; CDC2L3; CDC-related protein kinase p58; CDK11; CDK11 p110; CDK11 p46; CDK11 p58; CDK11A; Cdk11b; CDK11-p110; CDK11-p46; CDK11-p58; cell division cycle 2 homolog-like 1; cell division cycle 2 homolog-like 2; cell division cycle 2-like 1; cell division cycle 2-like 1 (PITSLRE proteins); cell division cycle 2-like 2; cell division cycle 2-like protein kinase 1; Cell division protein kinase 11; cell division protein kinase 11B; CLK-1; cyclin dependent kinase 11B; cyclin-dependent kinase 11; Cyclin-dependent kinase 11B; galactosyltransferase-associated protein kinase p58/GTA; p58; p58 CLK-1; p58CDC2L1; p58CLK-1; PITSLRE proteins; PITSLRE serine/threonine-protein kinase CDC2L1; PITSLREA; PK58 |
| Gene Symbols | CDK11B |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.